missing translation for 'onlineSavingsMsg'
Learn More

3-(Tri-n-butylstannyl)pyridine, 97%

Product Code. 15478077
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478077 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15478077 Supplier Thermo Scientific Alfa Aesar Supplier No. H51268.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 59020-10-9 | C17H31NSn | 368.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59020-10-9
Density 1.153
Molecular Formula C17H31NSn
MDL Number MFCD00052054
Quantity 1 g
UN Number UN2788
Synonym 3-tributylstannyl pyridine, 3-1,1,1-tributylstannyl pyridine, 3-pyridyltri-n-butyltin, pyridine, 3-tributylstannyl, 3-tri-n-butylstannyl pyridine, 3-pyridyltributylstannane, 3-tributylstannylpyridine, 3-tris but-1-ylstannyl pyridine, 3-tributylstannanyl-pyridine, 3-pyridyltributyltin
InChI Key CFQJBWKKHCMCGJ-UHFFFAOYSA-N
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CC=C1
IUPAC Name tributyl(pyridin-3-yl)stannane
Molecular Weight (g/mol) 368.152
PubChem CID 3483848
Formula Weight 368.15
Percent Purity 97%
Chemical Name or Material 3-(Tri-n-butylstannyl)pyridine
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.