missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethyl)crotonic acid, 96%

Product Code. 11366666
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11366666

Brand: Thermo Scientific Alfa Aesar L03816.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69056-67-3 | C5H5F3O2 | 154.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-67-3
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.09
MDL Number MFCD00014374,MFCD01457157
InChI Key QRRCTLYMABZQCS-NSCUHMNNSA-N
Synonym beta-trifluoromethylcrotonic acid, 3-trifluoromethylcrotonic acid, 3-trifluoromethyl crotonic acid, 2z-4,4,4-trifluoro-3-methylbut-2-enoic acid, heddpadiaicichiceiumlbbp, z-4,4,4-trifluoro-3-methylbut-2-enoic acid, 4,4,4-trifluoro-3-methyl-2-butenoic acid, 2-butenoic acid,4,4,4-trifluoro-3-methyl-, 2z
PubChem CID 5708835
IUPAC Name (Z)-4,4,4-trifluoro-3-methylbut-2-enoic acid
SMILES C\C(=C/C(O)=O)C(F)(F)F

Specifications

Melting Point 28°C to 30°C
Density 1.318
Boiling Point 158°C to 163°C
Flash Point 79°C (174°F)
Refractive Index 1.385
Quantity 25 g
UN Number UN3261
Beilstein 1769204
Formula Weight 154.09
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethyl)crotonic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.