missing translation for 'onlineSavingsMsg'
Learn More

3'-(Trifluoromethyl)propiophenone, 97%, Thermo Scientific™

Product Code. 11490681
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11490681

Brand: Thermo Scientific Alfa Aesar B20147.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1533-03-5
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.176
MDL Number MFCD00039227
InChI Key UBTPNKXHYILGJU-UHFFFAOYSA-N
Synonym 3'-trifluoromethyl propiophenone, m-trifluoromethylpropiophenone, 1-3-trifluoromethyl phenyl propan-1-one, 1-propanone, 1-3-trifluoromethyl phenyl, propiophenone, 3'-trifluoromethyl, 3-trifluoromethyl propiophenone, 1-3-trifluoromethyl-phenyl-propan-1-one, 1-3-trifluoromethyl phenyl-1-propanone
PubChem CID 137064
IUPAC Name 1-[3-(trifluoromethyl)phenyl]propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)C(F)(F)F

Specifications

Melting Point 18°C to 19°C
Density 1.202
Boiling Point 216°C to 217°C
Flash Point 108°C (226°F)
Refractive Index 1.4596
Quantity 5 g
Beilstein 2101539
Formula Weight 202.18
Percent Purity 97%
Chemical Name or Material 3'-(Trifluoromethyl)propiophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.