missing translation for 'onlineSavingsMsg'
Learn More

3-(Trimethylsilyl)propionic Acid-d4 Sodium Salt, TRC

Product Code. 30449541
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449541 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30449541 Supplier Toronto Research Chemicals Supplier No. TRCT7968821G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Trimethylsilyl)propionic Acid-d4 Sodium Salt
CAS 24493-21-8
Molecular Formula C6 2H4 H9 O2 Si . Na
Quantity 1 g
Synonym Sodium 3-(Trimethylsilyl)propionate-2,2,3,3-d4, 2,2,3,3-d4-3-(Trimethylsilyl)propionic Acid Sodium Salt
InChI Formula InChI=1S/C6H14O2Si.Na/c1-9(2,3)5-4-6(7)8;/h4-5H2,1-3H3,(H,7,8);/q;+1/p-1/i4D2,5D2;
SMILES [Na+].[2H]C([2H])(C(=O)[O-])C([2H])([2H])[Si](C)(C)C
IUPAC Name sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate
Molecular Weight (g/mol) 172.27
Formula Weight 172.0834
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.