missing translation for 'onlineSavingsMsg'
Learn More

3-Trimethylsilylpropynal, 97%

Product Code. 11470011
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470011 5 g 5g
11450011 0.25 g 0.25g
11460011 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470011 Supplier Thermo Scientific Alfa Aesar Supplier No. 044763.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2975-46-4 | C6H10OSi | 126.23 g/mol

3-Trimethylsilylpropynal acts as an important organic intermediate. It is used in agrochemicals, pharmaceuticals and dyestuff field. Further, it is used in the preparation of [3-(trimethylsilyl)-2-propinyliden]-malononitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2975-46-4
Molecular Formula C6H10OSi
Molecular Weight (g/mol) 126.23
MDL Number MFCD01103709
InChI Key LJRWLSKYGWLYIM-UHFFFAOYSA-N
Synonym 3-trimethylsilylpropynal, 3-trimethylsilyl-2-propynal, 3-trimethylsilyl propiolaldehyde, 3-trimethylsilyl propynal, trimethylsilyl propiolaldehyde, 2-propynal, 3-trimethylsilyl, trimethylsilylpropynal, 3-trimethylsilyl prop-2-ynal
PubChem CID 2760865
IUPAC Name 3-trimethylsilylprop-2-ynal
SMILES C[Si](C)(C)C#CC=O

Specifications

Color Colorless to Yellow
Density 0.86 g/mL
Boiling Point 58°C to 60°C (37.5 mmHg)
Flash Point 29°C (84°F)
Linear Formula (CH3)3SiC≡CCHO
Refractive Index 1.4435
Quantity 5 g
UN Number UN1993
Sensitivity Air and moisture sensitive
Formula Weight 126.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Trimethylsilylpropynal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.