missing translation for 'onlineSavingsMsg'
Learn More

3-((z)-Amino(hydroxyimino)methyl)benzoic acid, ≥90%, Thermo Scientific™

Product Code. 10300323
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10300323 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10300323 Supplier Thermo Scientific Maybridge Supplier No. MO07614DA
 Restricted Item
Estimated Shipment: 15-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 199447-10-4
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C8H8N2O3
MDL Number MFCD02083499
Quantity 1 g
Synonym 3-hydroxyamino iminomethyl-benzoic acid, 3-z-amino hydroxyimino methyl benzoic acid, 3-n'-hydroxycarbamimidoyl benzoic acid, 3-z-n'-hydroxycarbamimidoyl benzoic acid, 3-amino hydroxyimino methyl benzoic acid, benzoic acid, 3-n-hydroxycarbamimidoyl, benzoic acid, 3-z-amino hydroxyimino methyl, benzoic acid, 3-hydroxyamino iminomethyl-9ci
InChI Key LJCLPFBIVUPTAR-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)C(=O)O)C(=NO)N
IUPAC Name 3-[(Z)-N'-hydroxycarbamimidoyl]benzoic acid
Molecular Weight (g/mol) 180.163
PubChem CID 9603503
Formula Weight 180.16
Percent Purity 90%
Chemical Name or Material 3-[(Z)-amino(hydroxyimino)methyl]benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.