Learn More
TubeWriter 300 - Custom Rack Side Print for Any Labware
Product Code. 30067433
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
Product Code. Product Type Unit Size
30067433 Labeling Rack Each
1 options
Product Code. 30067433 Supplier TubeWriter Supplier No. PS0000XX300Custom
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Custom racks made to exact specifications to print directly on side of unique labware
  • Custom racks designed for 300
  • Racks made to your exact specifications to directly print and label on the side of any labware
  • Rack will ensure correct alignment and holds labware securely
  • No time-consuming sticky labels or handwriting required
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Product Type Labeling Rack