missing translation for 'onlineSavingsMsg'
Learn More

TubeWriter 300 - Standard Rack Side Print for 0.5 mL Microcentrifuge Tubes

Product Code. 30067419
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30067419 Labeling Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067419 Supplier TubeWriter Supplier No. PS0500MC300
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Standard rack for directly printing on the side of 0.5 mL microcentrifuge tubes

  • Standard printing rack designed for the 300
  • Prints and labels directly on the side of 0.5 mL microcentrifuge tubes
  • Rack ensures correct alignment and holds tubes securely
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions

Specifications

Product Type Labeling Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.