Learn More
TubeWriter 300 - Standard Rack Side Print for 50 mL Conical Centrifuge Tubes
Product Code. 30067413
Change view
Click to view available options
Product Type:
Labeling Rack
Unit Size:
Each
Product Code. Product Type Unit Size
30067413 Labeling Rack Each
1 options
Product Code. 30067413 Supplier TubeWriter Supplier No. PS0050CN300
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Standard rack for directly printing on the side of 50 mL conical centrifuge tubes
  • Standard printing rack designed for the 300
  • Prints and labels directly on the side of 50 mL conical centrifuge tubes
  • Rack ensures correct alignment and holds tubes securely
  • No time-consuming sticky labels or handwriting required
  • Printing an take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications
Product Type Labeling Rack