missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Methylcarbamate, TRC

Product Code. 30332585
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332585 Supplier Toronto Research Chemicals Supplier No. TRCB6940205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Methylcarbamate
CAS 16066-84-5
Molecular Formula C6H13NO2
Quantity 5 g
Synonym (tert-Butoxycarbonyl)methylamine, Methylcarbamic Acid tert-butyl Ester, N-(tert-Butoxycarbonyl)methylamine, N-Methylcarbamic Acid tert-Butyl Ester, tert-Butyl N-methylcarbamate, Methyl Carbamic Acid tert-Butyl Ester, Methyl Carbamic Acid 1,1-Dimethylethyl Ester, N-Methylcarbamic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C6H13NO2/c1-6(2,3)9-5(8)7-4/h1-4H3,(H,7,8)
SMILES CNC(=O)OC(C)(C)C
IUPAC Name tert-butyl N-methylcarbamate
Molecular Weight (g/mol) 131.17
Formula Weight 131.0946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.