missing translation for 'onlineSavingsMsg'
Learn More

O6-Benzyl-8-bromo-N9-[3’,5’-O-(1,1,3,3-tetrakis(isopropyl)-1,3-disiloxanediyl)-Beta-D-2’-deoxyribofuranosyl]guanine, TRC

Product Code. 30335878
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335878 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335878 Supplier Toronto Research Chemicals Supplier No. TRCB23345025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O6-Benzyl-8-bromo-N9-[3',5'-O-(1,1,3,3-tetrakis(isopropyl)-1,3-disiloxanediyl)-beta-D-2'-deoxyribofuranosyl]guanine
CAS 328394-26-9
Molecular Formula C29 H44 Br N5 O5 Si2
Quantity 25 mg
Synonym 8-Bromo-2'-deoxy-6-O-(phenylmethyl)-3',5'-O-[1,1,3,3-tetrakis(1-methylethyl)-1,3-disiloxanediyl]guanosine
InChI Formula InChI=1S/C29H44BrN5O5Si2/c1-17(2)41(18(3)4)37-16-23-22(39-42(40-41,19(5)6)20(7)8)14-24(38-23)35-26-25(32-28(35)30)27(34-29(31)33-26)36-15-21-12-10-9-11-13-21/h9-13,17-20,22-24H,14-16H2,1-8H3,(H2,31,33,34)/t22-,23+,24+/m0/s1
SMILES CC(C)[Si]1(OC[C@H]2O[C@H](C[C@@H]2O[Si](O1)(C(C)C)C(C)C)n3c(Br)nc4c(OCc5ccccc5)nc(N)nc34)C(C)C
IUPAC Name 9-[(6aR,8R,9aS)-2,2,4,4-tetra(propan-2-yl)-6a,8,9,9a-tetrahydro-6H-furo[3,2-f][1,3,5,2,4]trioxadisilocin-8-yl]-8-bromo-6-phenylmethoxypurin-2-amine
Molecular Weight (g/mol) 678.765
Formula Weight 677.206
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.