missing translation for 'onlineSavingsMsg'
Learn More

Bifenthrin, TRC

Product Code. 30339275
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30339275 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339275 Supplier Toronto Research Chemicals Supplier No. TRCB383000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bifenthrin
CAS 82657-04-3
Purity Grade Notes HPLC
Molecular Formula C23 H22 Cl F3 O2
Quantity 100 mg
Synonym Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (2-methyl[1,1'-biphenyl]-3-yl)methyl ester, [1α,3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (2-methyl[1,1'-biphenyl]-3-yl)methyl ester, (1R,3R)-rel- (9CI), (±)-Bifenthrin, AGST 02002, Bifenthrin, Bifenthrine, Bifenture, Biflex, Biflex FT, Biphenate, Biphenthrin, Biphentrin, Bixan, Bixan MG, Brigade, Brigade 10WP, Brigata Flo, Capture, Capture (pesticide), Capture 2EC, Capture LFR, Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (2-methyl[1,1'-biphenyl]-3-yl)methyl ester, [1α,3α(Z)]-, DeterMite, Discipline, Empower, FMC 54800, Fanfare, Gyro, Kiros EV, Onyx, Onyx (insecticide), Seizer, Semafor, Silencer, Sniper, Sniper (pyrethroid insecticide), Talstar, Talstar EZ, Talstar F, Talstar Pro, TalstarOne, rac-Bifenthrin
InChI Formula InChI=1S/C23H22ClF3O2/c1-14-16(10-7-11-17(14)15-8-5-4-6-9-15)13-29-21(28)20-18(22(20,2)3)12-19(24)23(25,26)27/h4-12,18,20H,13H2,1-3H3/b19-12-/t18-,20-/m1/s1
SMILES Cc1c(COC(=O)[C@H]2[C@@H](\C=C(/Cl)\C(F)(F)F)C2(C)C)cccc1c3ccccc3
IUPAC Name (2-methyl-3-phenylphenyl)methyl (1S,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 422.87
Formula Weight 422.126
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.