missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Calcium Sulfate |
| CAS | 7778-18-9 |
| Molecular Formula | CaSO4 |
| Quantity | 5 g |
| Synonym | A 30, A 30 (sulfate), Basic Calcium Sulfate, CA 5, CA 5 (Sulfate), CAB 30, CAS 20, CAS 20-4, Calcium Sulfate, Calcium Sulfate (CaSO4), Calcium Sulphate, Calmatrix, Capset, CoCoat P 80HB, Crysalba, D 1, D 1 (sulfate), D 101A, D 101A (sulfate), Denka Sigma 1000, Drierite, E 541, Franklin Fiber H 45, GR, GR (sulfate), HG 2, HG 2 (sulfate), JAX CS, KemWhite CG 80, NP-M 01, NP-MO 1, NP-S 02, Oparex 10, Osteoset, Raddichem 27, SK 10, SSS-A, Sulfuric acid Calcium Salt, Sulfuric Acid Calcium(2+) Salt (1:1), SurgiPlaster, TR 100 (Sulfate), Thiolite |
| InChI Formula | InChI=1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2 |
| SMILES | [Ca+2].[O-]S(=O)(=O)[O-] |
| IUPAC Name | calcium;sulfate |
| Molecular Weight (g/mol) | 136.14 |
| Formula Weight | 135.9143 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?