missing translation for 'onlineSavingsMsg'
Learn More

Azulene, TRC

Product Code. 30342540
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30342540

Brand: Toronto Research Chemicals TRCA965250100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azulene
CAS 275-51-4
Molecular Formula C10 H8
Quantity 100 mg
Synonym Azulene (6CI, 8CI, 9CI, ACI), Bicyclo[5.3.0]decapentaene, Cyclopentacycloheptene, NSC 89248
InChI Formula InChI=1S/C10H8/c1-2-5-9-7-4-8-10(9)6-3-1/h1-8H
SMILES c1ccc2cccc2cc1
IUPAC Name azulene
Molecular Weight (g/mol) 128.17
Formula Weight 128.0626
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.