missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine, TRC

Product Code. 30343615
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343615 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343615 Supplier Toronto Research Chemicals Supplier No. TRCB414053100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzydamine
CAS 642-72-8
Molecular Formula C19H23N3O
Quantity 100 mg
Synonym 1-Propanamine, N,N-dimethyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-, 1H-Indazole, 1-benzyl-3-[3-(dimethylamino)propoxy]- (7CI,8CI), N,N-Dimethyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine, 1-Benzyl-3-(γ-dimethylaminopropoxy)-1H-indazole, 1-Benzyl-3-[3-(dimethylamino)propoxy]-1H-indazole, 1-Benzyl-3-[3-(dimethylamino)propoxy]indazole, 1-Benzyl-3-dimethylaminopropoxy-1H-indazole, Benzidamine, Benzindamine, Benzydamine
InChI Formula InChI=1S/C19H23N3O/c1-21(2)13-8-14-23-19-17-11-6-7-12-18(17)22(20-19)15-16-9-4-3-5-10-16/h3-7,9-12H,8,13-15H2,1-2H3
SMILES CN(C)CCCOc1nn(Cc2ccccc2)c3ccccc13
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine
Molecular Weight (g/mol) 309.4
Formula Weight 309.1841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.