missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bifenthrin, TRC
Specifications
Specifications
| Chemical Name or Material | Bifenthrin |
| CAS | 82657-04-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 H22 Cl F3 O2 |
| Quantity | 1 g |
| Synonym | Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (2-methyl[1,1'-biphenyl]-3-yl)methyl ester, [1α,3α(Z)]-(±)-, Cyclopropanecarboxylic acid, 3-[(1Z)-2-chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethyl-, (2-methyl[1,1'-biphenyl]-3-yl)methyl ester, (1R,3R)-rel- (9CI), (±)-Bifenthrin, AGST 02002, Bifenthrin, Bifenthrine, Bifenture, Biflex, Biflex FT, Biphenate, Biphenthrin, Biphentrin, Bixan, Bixan MG, Brigade, Brigade 10WP, Brigata Flo, Capture, Capture (pesticide), Capture 2EC, Capture LFR, Cyclopropanecarboxylic acid, 3-(2-chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethyl-, (2-methyl[1,1'-biphenyl]-3-yl)methyl ester, [1α,3α(Z)]-, DeterMite, Discipline, Empower, FMC 54800, Fanfare, Gyro, Kiros EV, Onyx, Onyx (insecticide), Seizer, Semafor, Silencer, Sniper, Sniper (pyrethroid insecticide), Talstar, Talstar EZ, Talstar F, Talstar Pro, TalstarOne, rac-Bifenthrin |
| InChI Formula | InChI=1S/C23H22ClF3O2/c1-14-16(10-7-11-17(14)15-8-5-4-6-9-15)13-29-21(28)20-18(22(20,2)3)12-19(24)23(25,26)27/h4-12,18,20H,13H2,1-3H3/b19-12-/t18-,20-/m1/s1 |
| SMILES | Cc1c(COC(=O)[C@H]2[C@@H](\C=C(/Cl)\C(F)(F)F)C2(C)C)cccc1c3ccccc3 |
| IUPAC Name | (2-methyl-3-phenylphenyl)methyl (1S,3S)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate |
| Molecular Weight (g/mol) | 422.87 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?