missing translation for 'onlineSavingsMsg'
Learn More

(R)-Azelastine N-Oxide (Mixture of Diastereomers), TRC

Product Code. 30345280
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345280 10 mg 10mg
30337165 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30345280 Supplier Toronto Research Chemicals Supplier No. TRCA80826510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Azelastine N-oxide
CAS 1346617-18-2
Molecular Formula C22 H24 Cl N3 O2
Quantity 10 mg
Synonym 4-[(4-Chlorophenyl)methyl]-2-[(4R)-hexahydro-1-methyl-1-oxido-1H-azepin-4-yl]-1(2H)-phthalazinone, (R)-Azelastine N-Oxide
InChI Formula InChI=1S/C22H24ClN3O2/c1-26(28)13-4-5-18(12-14-26)25-22(27)20-7-3-2-6-19(20)21(24-25)15-16-8-10-17(23)11-9-16/h2-3,6-11,18H,4-5,12-15H2,1H3/t18-,26?/m1/s1
SMILES C[N+]1([O-])CCC[C@H](CC1)N2N=C(Cc3ccc(Cl)cc3)c4ccccc4C2=O
IUPAC Name 4-[(4-chlorophenyl)methyl]-2-[(4R)-1-methyl-1-oxidoazepan-1-ium-4-yl]phthalazin-1-one
Molecular Weight (g/mol) 397.9
Formula Weight 397.1557
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.