missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apamin TFA Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Apamin TFA Salt |
| Molecular Formula | C79H131N31O24S4 . x(C2HF3O2) |
| Quantity | 1 mg |
| Synonym | Apamin TFA Salt, 32,15-(Iminoethaniminopropanodithiomethano)-31H-pyrrolo[2,1-e1][1,2,5,8,11,14,17,20,23,26,29,32,35,38]dithiadodecaazacyclohentetracontine Cyclic Peptide Derivative xTFA Salt, Apamin (reduced) Cyclic (1→11),(3→15)-Bis(disulfide) xTFA Salt, Apamine xTFA Salt, L-Cysteinyl-L-asparaginyl-L-cysteinyl-L-lysyl-L-alanyl-L-prolyl-L-α-glutamyl-L-threonyl-L-alanyl-L-leucyl-L-cysteinyl-L-alanyl-L-arginyl-L-arginyl-L-cysteinyl-L-glutaminyl-L-glutaminyl-L-histidinamide cyclic (1→11),(3→15)-bis(disulfide) xTFA Salt, Ro 23-6721 xTFA Salt |
| InChI Formula | InChI=1S/C79H131N31O24S4.C2HF3O2/c1-35(2)26-49-71(128)105-51-31-136-135-30-41(81)64(121)104-50(28-57(84)114)72(129)106-52(73(130)98-42(12-7-8-22-80)66(123)96-38(5)77(134)110-25-11-15-54(110)75(132)101-46(18-21-58(115)116)69(126)107-59(39(6)111)76(133)95-37(4)63(120)103-49)32-137-138-33-53(109-108-47(14-10-24-92-79(88)89)70(127)97-43(13-9-23-91-78(86)87)65(122)94-36(3)60(117)61(51)118)74(131)100-44(16-19-55(82)112)67(124)99-45(17-20-56(83)113)68(125)102-48(62(85)119)27-40-29-90-34-93-40;3-2(4,5)1(6)7/h29,34-39,41-54,59,108-109,111H,7-28,30-33,80-81H2,1-6H3,(H2,82,112)(H2,83,113)(H2,84,114)(H2,85,119)(H,90,93)(H,94,122)(H,95,133)(H,96,123)(H,97,127)(H,98,130)(H,99,124)(H,100,131)(H,101,132)(H,102,125)(H,103,120)(H,104,121)(H,105,128)(H,106,129)(H,107,126)(H,115,116)(H4,86,87,91)(H4,88,89,92);(H,6,7)/t36-,37-,38-,39+,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,59-;/m0./s1 |
| SMILES | N[C@@H](CSSC[C@H](NC([C@H](CC(C)C)NC([C@@H](N1)C)=O)=O)C(C([C@@H](NC([C@H](CCCNC(N)=N)N2)=O)C)=O)=O)C(N[C@@H](CC(N)=O)C(N[C@@H](CSSC[C@H](NN[C@@H](CCCNC(N)=N)C2=O)C(N[C@@H](CCC(N)=O)C(N[C@@H](CCC(N)=O)C(N[C@H](C(N)=O)CC3=CN=CN3)=O)=O)=O)C(N[C@@H](CCCCN)C(N[C@H](C(N4CCC[C@H]4C(N[C@@H](CCC(O)=O)C(N[C@@H]([C@H](O)C)C1=O)=O)=O)=O)C)=O)=O)=O)=O.OC(C(F)(F)F)=O |
| IUPAC Name | 3-((3S,6S,9S,12S,15R,18S,21S,24S,27R,32R,35S,38S,43aS,46S,49R)-49-amino-27-(((S)-5-amino-1-(((S)-5-amino-1-(((S)-1-amino-3-(1H-imidazol-5-yl)-1-oxopropan-2-yl)amino)-1,5-dioxopentan-2-yl)amino)-1,5-dioxopentan-2-yl)carbamoyl)-46-(2-amino-2-oxoethyl)-35-(4-aminobutyl)-21,24-bis(3-guanidinopropyl)-6-((R)-1-hydroxyethyl)-12-isobutyl-9,18,38-trimethyl-1,4,7,10,11,16,19,20,23,33,36,39,45,48-tetradecaoxotetracontahydro-31H-32,15-(epiminoethanoiminopropanodithiomethano)pyrrolo[2,1-e1][1,2]dithia[5,6,9,14,17,18,23,26,29,32,35,38]dodecaazacyclohentetracontin-3-yl)propanoic acid TFA Salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?