missing translation for 'onlineSavingsMsg'
Learn More

Boldenone Propionate, TRC

Product Code. 30347519
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347519 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347519 Supplier Toronto Research Chemicals Supplier No. TRCB67516050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Oxoandrosta-1,4-dien-17beta-yl Propanoate
CAS 977-32-2
Purity Grade Notes HPLC
Molecular Formula C22 H30 O3
Quantity 50 mg
Synonym 3-Oxoandrosta-1,4-dien-17β-yl propanoate, Androsta-1,4-dien-3-one, 17-(1-oxopropoxy)-, (17β)-, Androsta-1,4-dien-3-one, 17β-hydroxy-, propionate (6CI,7CI,8CI), (17β)-17-(1-Oxopropoxy)androsta-1,4-dien-3-one, Boldenone propionate
InChI Formula InChI=1S/C22H30O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h9,11,13,16-19H,4-8,10,12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1
SMILES CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 342.47
Formula Weight 342.2195
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.