missing translation for 'onlineSavingsMsg'
Learn More

4'-Chlorobiphenyl-2-ylamine, TRC

Product Code. 30349094
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30349094 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349094 Supplier Toronto Research Chemicals Supplier No. TRCC364305250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Chlorobiphenyl-2-ylamine
CAS 1204-44-0
Molecular Formula C12H10ClN
Quantity 250 mg
Synonym 4'-Chloro-[1,1'-biphenyl]-2-amine, 4'-Chloro-2-biphenylamine, 2-Amino-4'-chlorobiphenyl, 4'-Chloro-1,1'-biphenyl-2-amine, NSC 95712
InChI Formula InChI=1S/C12H10ClN/c13-10-7-5-9(6-8-10)11-3-1-2-4-12(11)14/h1-8H,14H2
SMILES ClC1=CC=C(C2=C(N)C=CC=C2)C=C1
IUPAC Name 4'-chloro-[1,1'-biphenyl]-2-amine
Molecular Weight (g/mol) 203.67
Formula Weight 203.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.