missing translation for 'onlineSavingsMsg'
Learn More

Dansyl Ethylenediamine, TRC

Product Code. 30349219
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349219 100 mg 100mg
30360013 500 mg 500mg
30366671 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30349219 Supplier Toronto Research Chemicals Supplier No. TRCD179500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35060-08-3
Molecular Formula C14 H19 N3 O2 S
Molecular Weight (g/mol) 293.38
IUPAC Name N-(2-aminoethyl)-5-(dimethylamino)naphthalene-1-sulfonamide
SMILES CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCN

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H19N3O2S/c1-17(2)13-7-3-6-12-11(13)5-4-8-14(12)20(18,19)16-10-9-15/h3-8,16H,9-10,15H2,1-2H3
Formula Weight 293.1198
Chemical Name or Material Dansyl Ethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.