missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbuterol Hemisulfate Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Carbuterol Hemisulfate Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | 2 C13 H21 N3 O3 . H2 O4 S |
| Quantity | 2.5 mg |
| Synonym | Carbuterol Hemisulfate, Urea, N-[5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]-, sulfate (2:1), N-[5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-2-hydroxyphenyl]urea Hemisulfate, Bronsecur, Carbuterol Hemisulfate Pirem, SKF 40383 Hemisulfate, SKF 40383A Hemisulfate |
| InChI Formula | InChI=1S/2C13H21N3O3.H2O4S/c2*1-13(2,3)15-7-11(18)8-4-5-10(17)9(6-8)16-12(14)19;1-5(2,3)4/h2*4-6,11,15,17-18H,7H2,1-3H3,(H3,14,16,19);(H2,1,2,3,4) |
| SMILES | CC(C)(C)NCC(O)c1ccc(O)c(NC(=O)N)c1.CC(C)(C)NCC(O)c2ccc(O)c(NC(=O)N)c2.OS(=O)(=O)O |
| IUPAC Name | [5-[2-(tert-butylamino)-1-hydroxyethyl]-2-hydroxyphenyl]urea;sulfuric acid |
| Molecular Weight (g/mol) | 632.7268 |
| Formula Weight | 632.284 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?