missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Etoricoxib-d4, TRC

Product Code. 30349482
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349482 10 mg 10mg
30352198 100 mg 100mg
30355746 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30349482 Supplier Toronto Research Chemicals Supplier No. TRCD29689710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethyl Etoricoxib-d4
Molecular Formula C17 D4 H9 Cl N2 O2 S
Quantity 10 mg
Synonym 5-Chloro-3-(4-methylsulfonylphenyl)-2-(3-pyridyl)pyridine-d4
InChI Formula InChI=1S/C17H13ClN2O2S/c1-23(21,22)15-6-4-12(5-7-15)16-9-14(18)11-20-17(16)13-3-2-8-19-10-13/h2-11H,1H3/i2D,3D,8D,10D
SMILES [2H]c1nc([2H])c(c([2H])c1[2H])c2ncc(Cl)cc2c3ccc(cc3)S(=O)(=O)C
IUPAC Name 3-[5-chloro-3-(4-methylsulfonylphenyl)pyridin-2-yl]-2,4,5,6-tetradeuteriopyridine
Molecular Weight (g/mol) 348.84
Formula Weight 348.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.