missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentyl-4-nitrosopiperazine-d9, TRC

Product Code. 30349748
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349748 2.5 mg 2.5mg
30364514 25 mg -
30360026 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30349748 Supplier Toronto Research Chemicals Supplier No. TRCC2137972.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C9 D9 H8 N3 O
Molecular Weight (g/mol) 192.306
IUPAC Name 1-nitroso-4-(1,2,2,3,3,4,4,5,5-nonadeuteriocyclopentyl)piperazine
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(N2CCN(CC2)N=O)C1([2H])[2H]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C9H17N3O/c13-10-12-7-5-11(6-8-12)9-3-1-2-4-9/h9H,1-8H2/i1D2,2D2,3D2,4D2,9D
Formula Weight 192.194
Chemical Name or Material 1-Cyclopentyl-4-nitrosopiperazine-d9
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.