missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) Chloride Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Cobalt(II) Chloride Hydrate |
| CAS | 69098-14-2 |
| Molecular Formula | Cl2Co •xH2O |
| Quantity | 5 g |
| Synonym | Cobalt Chloride Hydrate, Cobalt Dichloride Hydrate, Cobalt Dichloride Hydrate (CoCl2), Cobalt(2+) Chloride Hydrate, Cobalt(II) Chloride Hydrate, Cobaltous Chloride Hydrate, Cobaltous Dichloride Hydrate, Dichlorocobalt Hydrate, NSC 51149 |
| InChI Formula | InChI=1S/2ClH.Co.H2O/h2*1H;;1H2/q;;+2;/p-2 |
| SMILES | Cl[Co]Cl |
| IUPAC Name | dichlorocobalt;hydrate |
| Molecular Weight (g/mol) | 129.841801 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?