missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldithiocarbamic Acid Sodium Salt Dihydrate, TRC

Product Code. 30350923
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350923 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350923 Supplier Toronto Research Chemicals Supplier No. TRCD4137502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethyldithiocarbamate Sodium Dihydrate
CAS 72140-17-1
Molecular Formula C3 H6 N S2 . 2 H2 O . Na
Quantity 2.5 g
Synonym Dimethyldithiocarbamic Acid Sodium Salt Dihydrate, Carbamodithioic acid, dimethyl-, sodium salt, dihydrate (9CI), Sodium dimethyldithiocarbamate dihydrate, N,N-Dimethyldithiocarbamate Sodium Dihydrate
InChI Formula InChI=1S/C3H7NS2.Na.2H2O/c1-4(2)3(5)6;;;/h1-2H3,(H,5,6);;2*1H2/q;+1;;/p-1
SMILES O.O.[Na+].CN(C)C(=S)[S-]
IUPAC Name sodium;N,N-dimethylcarbamodithioate;dihydrate
Molecular Weight (g/mol) 179.237
Formula Weight 179.005
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.