missing translation for 'onlineSavingsMsg'
Learn More

(R)-Hydroxychloroquine, TRC

Product Code. 30350959
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350959 100 mg 100mg
30368165 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350959 Supplier Toronto Research Chemicals Supplier No. TRCC373710100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137433-23-9
Molecular Formula C18 H26 Cl N3 O
Molecular Weight (g/mol) 335.87
Synonym Ethanol, 2-[[4-[(7-Chloro-4-Quinolinyl)Amino]Pentyl]Ethylamino]-, (R)-, 2-[[(4R)-4-[(7-Chloro-4-Quinolinyl)Amino]Pentyl]Ethylamino]Ethanol, 2-[[(4R)-4-[(7-Chloro-4-Quinolinyl)Amino]Pentyl]Ethylamino]-Ethanol
IUPAC Name 2-[[(4R)-4-[(7-chloroquinolin-4-yl)amino]pentyl]-ethylamino]ethanol
SMILES Ccn(Cco)Ccc[C@@H](C)Nc1Ccnc2Cc(Cl)Ccc12

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H26ClN3O/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21)/t14-/m1/s1
Formula Weight 335.1764
Chemical Name or Material (R)-Hydroxychloroquine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.