missing translation for 'onlineSavingsMsg'
Learn More

2-[2-[(5-carboxypentyl)amino]-2-oxoethyl]-2-hydroxybutanedioic Acid, TRC

Product Code. 30351437
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351437 50 mg 50mg
30350531 10 mg 10mg
30358206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351437 Supplier Toronto Research Chemicals Supplier No. TRCC18117550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1675217-42-1
Molecular Formula C12H19NO8
Molecular Weight (g/mol) 305.28
Synonym 2-[2-(5-Carboxypentylamino)-2-oxoethyl]malic Acid, 2-(2-((5-Carboxypentyl)amino)-2-oxoethyl)-2-hydroxysuccinic Acid, 3-Hydroxy-3,4-dicarboxy-butanamide-N-hexanoic Acid
IUPAC Name 2-[2-(5-carboxypentylamino)-2-oxoethyl]-2-hydroxybutanedioic acid
SMILES OC(=O)CCCCCNC(=O)CC(O)(CC(=O)O)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H19NO8/c14-8(13-5-3-1-2-4-9(15)16)6-12(21,11(19)20)7-10(17)18/h21H,1-7H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)
Formula Weight 305.1111
Chemical Name or Material 2-[2-[(5-carboxypentyl)amino]-2-oxoethyl]-2-hydroxybutanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.