missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetylurea-13C,15N2, TRC

Product Code. 30351507
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351507 25 mg 25mg
30354275 100 mg 100mg
30356811 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351507 Supplier Toronto Research Chemicals Supplier No. TRCC98964725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C313CH5N15N2O2
Molecular Weight (g/mol) 130.08
Synonym (2-Cyanoethanoyl)urea-13C,15N2, (Cyanoacetyl)urea-13C,15N2, NSC 6287-13C,15N2, N-(Aminocarbonyl)-2-cyanoacetamide-13C,15N2
IUPAC Name N-(carbamoyl-13C-15N)-2-cyanoacetamide-15N
SMILES [15NH2][13C]([15NH]C(CC#N)=O)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C4H5N3O2/c5-2-1-3(8)7-4(6)9/h1H2,(H3,6,7,8,9)/i4+1,6+1,7+1
Formula Weight 130.04
Chemical Name or Material Cyanoacetylurea-13C,15N2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.