missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Deoxy-2-chloro-D-glucose, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-chloro-2-deoxy-D-glucose |
| CAS | 14685-79-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6 H11 Cl O5 |
| Quantity | 10 mg |
| Synonym | D-Glucose, 2-chloro-2-deoxy-, 2-Chloro-2-deoxy-D-glucose, 2-Deoxy-2-chloro-D-glucose |
| InChI Formula | InChI=1S/C6H11ClO5/c7-3(1-8)5(11)6(12)4(10)2-9/h1,3-6,9-12H,2H2/t3-,4+,5+,6+/m0/s1 |
| SMILES | OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](Cl)C=O |
| IUPAC Name | (2R,3S,4R,5R)-2-chloro-3,4,5,6-tetrahydroxyhexanal |
| Molecular Weight (g/mol) | 198.6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction