missing translation for 'onlineSavingsMsg'
Learn More

Deltaline, TRC

Product Code. 30351802
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351802 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351802 Supplier Toronto Research Chemicals Supplier No. TRCD23166510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Deltaline
CAS 6836-11-9
Molecular Formula C27H41NO8
Quantity 10 mg
Synonym Eldeline, Delphelatine, Deltamine 6-Acetate, Eldelin, (1alpha,6beta,14alpha,16beta)-20-Ethyl-1,14,16-trimethoxy-4-methyl-7,8-[methylenebis(oxy)]-aconitane-6,10-diol 6-Acetate
InChI Formula InChI=1S/C27H41NO8/c1-7-28-12-23(3)9-8-17(32-5)26-20(23)21(36-14(2)29)27(22(26)28)25(34-13-35-27)11-16(31-4)15-10-24(26,30)19(25)18(15)33-6/h15-22,30H,7-13H2,1-6H3
SMILES CCN1CC2(C)CCC(OC)C34C2C(OC(=O)C)C5(OCOC56CC(OC)C7CC3(O)C6C7OC)C14
Molecular Weight (g/mol) 507.62
Formula Weight 507.2832
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.