missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-3'-Deoxythymidine, TRC

Product Code. 30352069
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30352069 25 mg
30358718 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352069 Supplier Toronto Research Chemicals Supplier No. TRCC36557525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25526-94-7
Molecular Formula C10 H13 Cl N2 O4
Molecular Weight (g/mol) 260.67
Synonym 1-(3-Chloro-2,3-dideoxy-beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione, Zidovudine Imp. B (EP), 3'-chloro-3'-deoxythymidine
IUPAC Name 1-[(2R,4S,5R)-4-chloro-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2C[C@H](Cl)[C@@H](CO)O2)C(=O)NC1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13ClN2O4/c1-5-3-13(10(16)12-9(5)15)8-2-6(11)7(4-14)17-8/h3,6-8,14H,2,4H2,1H3,(H,12,15,16)/t6-,7+,8+/m0/s1
Formula Weight 260.0564
Chemical Name or Material 1-(3-Chloro-2,3-dideoxy-Beta-D-erythro-pentofuranosyl)-5-methylpyrimidine-2,4(1H,3H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.