missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytarabine 5’-Monophosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Cytarabine 5'-Monophosphate |
| CAS | 7075-11-8 |
| Molecular Formula | C9 H14 N3 O8 P |
| Quantity | 100 mg |
| Synonym | 2(1H)-Pyrimidinone, 4-amino-1-(5-O-phosphono-β-D-arabinofuranosyl)-, Cytosine, 1-β-D-arabinofuranosyl-, 5'-(dihydrogen phosphate) (7CI,8CI), 4-Amino-1-(5-O-phosphono-β-D-arabinofuranosyl)-2(1H)-pyrimidinone, 1-β-D-Arabinofuranosylcytosine 5'-monophosphate, 1-β-D-Arabinofuranosylcytosine 5'-phosphate, Aracytidine 5'-monophosphate, Aracytidine 5'-phosphate, Cytosine arabinoside 5'-phosphate, Cytosine arabinoside monophosphate, Cytosine β-D-arabinoside 5'-monophosphate, Cytosine-β-D-arabinofuranoside-5'-monophosphoric acid, NSC 99445, Cytarabine 5'-monophosphate |
| InChI Formula | InChI=1S/C9H14N3O8P/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)/t4-,6-,7+,8-/m1/s1 |
| SMILES | NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(=O)(O)O)[C@@H](O)[C@@H]2O |
| IUPAC Name | [(2R,3S,4S,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate |
| Molecular Weight (g/mol) | 323.2 |
| Formula Weight | 323.0519 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?