missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromercuribenzoic Acid, TRC

Product Code. 30353134
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353134 2.5 g 2.5g
30367106 5 g -
30362979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353134 Supplier Toronto Research Chemicals Supplier No. TRCC3678002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-85-8
Molecular Formula C7 H5 Cl Hg O2
Molecular Weight (g/mol) 357.16
IUPAC Name (4-carboxyphenyl)-chloromercury
SMILES OC(=O)c1ccc([Hg]Cl)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C7H5O2.ClH.Hg/c8-7(9)6-4-2-1-3-5-6;;/h2-5H,(H,8,9);1H;/q;;+1/p-1
Formula Weight 357.9684
Chemical Name or Material 4-Chloromercuribenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.