missing translation for 'onlineSavingsMsg'
Learn More

Deferasirox Methyl Ester-d3, TRC

Product Code. 30353392
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353392 2.5 mg 2.5mg
30351982 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30353392 Supplier Toronto Research Chemicals Supplier No. TRCD2286772.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22H14D3N3O4
Molecular Weight (g/mol) 390.41
Synonym 4-[3,5-Bis(2-hydroxyphenyl)-1H-1,2,4-triazol-1-yl]-benzoic Acid Methyl Ester-d3
IUPAC Name trideuteriomethyl 4-[3,5-bis(2-hydroxyphenyl)-1,2,4-triazol-1-yl]benzoate
SMILES OC1=CC=CC=C1C2=NN(C3=CC=C(C(OC([2H])([2H])[2H])=O)C=C3)C(C4=C(O)C=CC=C4)=N2

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H17N3O4/c1-29-22(28)14-10-12-15(13-11-14)25-21(17-7-3-5-9-19(17)27)23-20(24-25)16-6-2-4-8-18(16)26/h2-13,26-27H,1H3/i1D3
Chemical Name or Material Deferasirox Methyl Ester-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.