missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroimidazo[1,2-a]pyridine-6-carbonitrile, TRC

Product Code. 30353666
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353666 2.5 mg 2.5mg
30349122 5 mg -
30360793 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353666 Supplier Toronto Research Chemicals Supplier No. TRCC3829302.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019020-03-1
Molecular Formula C8H4N3Cl
Molecular Weight (g/mol) 177.59
IUPAC Name 2-chloroimidazo[1,2-a]pyridine-6-carbonitrile
SMILES Clc1cn2cc(ccc2n1)C#N

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C8H4ClN3/c9-7-5-12-4-6(3-10)1-2-8(12)11-7/h1-2,4-5H
Formula Weight 177.0094
Chemical Name or Material 2-Chloroimidazo[1,2-a]pyridine-6-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.