missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(3-chloro-2-methylphenyl)nicotinamide, TRC

Product Code. 30353697
Change view
Click to view available options
Quantity:
100 mg
250 mg
2.5 g
Unit Size:
100mg
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353697 2.5 g 2.5g
30361903 250 mg 250mg
30359859 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353697 Supplier Toronto Research Chemicals Supplier No. TRCC3654802.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57841-61-9
Molecular Formula C13 H10 Cl2 N2 O
Molecular Weight (g/mol) 281.14
Synonym 2-Chloro-N-(3-chloro-2-methylphenyl)-3-pyridinecarboxamide
IUPAC Name 2-chloro-N-(3-chloro-2-methylphenyl)pyridine-3-carboxamide
SMILES Cc1c(Cl)cccc1NC(=O)c2cccnc2Cl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C13H10Cl2N2O/c1-8-10(14)5-2-6-11(8)17-13(18)9-4-3-7-16-12(9)15/h2-7H,1H3,(H,17,18)
Formula Weight 280.017
Chemical Name or Material 2-Chloro-N-(3-chloro-2-methylphenyl)nicotinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.