missing translation for 'onlineSavingsMsg'
Learn More

N-Desmethyl Rosuvastatin-d6 Disodium Salt, TRC

Product Code. 30353761
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353761 10 mg 10mg
30358720 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30353761 Supplier Toronto Research Chemicals Supplier No. TRCD29209210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21H18D6FN3Na2O6S
Molecular Weight (g/mol) 517.51
Synonym (E)-7-[4-(4-Fluorophenyl)-6-(isopropyl-d6)-2-mesylaminopyrimidin-5-yl]-(3R,5S)-dihydroxyhept-6-enoic Acid Disodium Salt
SMILES FC1=CC=C(C2=NC([N-]S(=O)(C)=O)=NC(C(C([2H])([2H])[2H])C([2H])([2H])[2H])=C2/C=C/[C@H](O)C[C@H](O)CC([O-])=O)C=C1.[Na+].[Na+]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H26FN3O6S.2Na/c1-12(2)19-17(9-8-15(26)10-16(27)11-18(28)29)20(13-4-6-14(22)7-5-13)24-21(23-19)25-32(3,30)31;;/h4-9,12,15-16,26-27H,10-11H2,1-3H3,(H2,23,24,25,28,29);;/q;2*+1/p-2/b9-8+;;/t15-,16-;;/m0../s1/i1D3,2D3;;
Percent Purity >95%
Chemical Name or Material N-Desmethyl Rosuvastatin-d6 Disodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.