missing translation for 'onlineSavingsMsg'
Learn More

Copper Oxychloride, TRC

Product Code. 30354310
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354310 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354310 Supplier Toronto Research Chemicals Supplier No. TRCC68538550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper Oxychloride
CAS 1332-65-6
Molecular Formula Cl Cu2 H3 O3
Quantity 50 g
InChI Formula InChI=1S/ClH.2Cu.3H2O/h1H;;;3*1H2/q;;+4;;;/p-4
SMILES O[Cu](O)(O)(Cl)[Cu]
IUPAC Name chloro-cuprio-trihydroxy-copper
Molecular Weight (g/mol) 213.57
Formula Weight 211.8363
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.