missing translation for 'onlineSavingsMsg'
Learn More

Cyclodecanone, TRC

Product Code. 30354579
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354579 500 mg 500mg
30354115 1 g 1g
30358368 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354579 Supplier Toronto Research Chemicals Supplier No. TRCC989098500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1502-06-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
IUPAC Name cyclodecanone
SMILES O=C1CCCCCCCCC1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H18O/c11-10-8-6-4-2-1-3-5-7-9-10/h1-9H2
Formula Weight 154.1358
Chemical Name or Material Cyclodecanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.