missing translation for 'onlineSavingsMsg'
Learn More

Desmethylene Oxobexarotene-13C4 Methyl Ester, TRC

Product Code. 30355305
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355305 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355305 Supplier Toronto Research Chemicals Supplier No. TRCD2911371MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethylene Oxobexarotene-13C4 Methyl Ester
CAS 1391054-72-0
Molecular Formula C2013C4H28O3
Quantity 1 mg
Synonym Methyl 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]benzoate -13C4, 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]-benzoic Acid Methyl Ester -13C4
InChI Formula InChI=1S/C24H28O3/c1-15-13-19-20(24(4,5)12-11-23(19,2)3)14-18(15)21(25)16-7-9-17(10-8-16)22(26)27-6/h7-10,13-14H,11-12H2,1-6H3/i2+1,3+1,4+1,5+1
SMILES [13CH3]C1([13CH3])CCC([13CH3])([13CH3])c2cc(C(=O)c3ccc(cc3)C(=O)OC)c(C)cc12
IUPAC Name methyl 4-[3-methyl-5,5,8,8-tetra((1^{13}C)methyl)-6,7-dihydronaphthalene-2-carbonyl]benzoate
Molecular Weight (g/mol) 368.45
Formula Weight 368.2173
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.