missing translation for 'onlineSavingsMsg'
Learn More

Cholic Acid Methyl Ester, TRC

Product Code. 30355422
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30355422 1 g 1g
30364653 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30355422 Supplier Toronto Research Chemicals Supplier No. TRCC4326101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448-36-8
Molecular Formula C25 H42 O5
Molecular Weight (g/mol) 422.6
Synonym Cholan-24-oic acid, 3,7,12-trihydroxy-, methyl ester, (3α,5β,7α,12α)-, Cholic acid, methyl ester (6CI,7CI,8CI), Methyl 3α,7α,12α-trihydroxy-5β-cholan-24-ate, Methyl 3α,7α,12α-trihydroxy-5β-cholan-24-oate, Methyl 3α,7α,12α-trihydroxy-5β-cholanoate, Methyl cholate, NSC 126794
IUPAC Name methyl (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H42O5/c1-14(5-8-22(29)30-4)17-6-7-18-23-19(13-21(28)25(17,18)3)24(2)10-9-16(26)11-15(24)12-20(23)27/h14-21,23,26-28H,5-13H2,1-4H3/t14-,15+,16-,17-,18+,19+,20-,21+,23+,24+,25-/m1/s1
Formula Weight 422.3032
Chemical Name or Material Cholic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.