missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Trifluoromethanesulfonate, TRC

Product Code. 30356329
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356329 10 g 10g
30358013 25 g 25g
30349947 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30356329 Supplier Toronto Research Chemicals Supplier No. TRCC68537510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34946-82-2
Molecular Formula C2CuF6O6S2
Molecular Weight (g/mol) 361.68
Synonym 1,1,1-Trifluoromethanesulfonic Acid Copper(2+) Salt (2:1), Trifluoromethanesulfonic Acid Copper(2+) Salt, Bis(trifluoromethanesulfonato)copper, Copper Bis(trifluoromethanesulfonate), Copper Ditriflate, Copper(2+) Bis(trifluoromethanesulfonate), Copper(2+) Triflate, Copper(2+) Trifluoromethylsulfonate, Copper(II) Triflate, Cupric Bis(trifluoromethanesulfonate), Cupric Triflate, Cupric Trifluoromethanesulfonate, Cupric Trifluoromethylsulfonate, Trifluoromethanesulfonic Acid Copper Salt (2:1), Trifluoromethanesulfonic Acid Copper(2+) Salt
IUPAC Name copper;trifluoromethanesulfonate
SMILES [Cu+2].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 10 g
InChI Formula InChI=1S/2CHF3O3S.Cu/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2
Formula Weight 360.8336
Chemical Name or Material Copper(II) Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.