missing translation for 'onlineSavingsMsg'
Learn More

Carboxine, TRC

Product Code. 30356401
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356401 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356401 Supplier Toronto Research Chemicals Supplier No. TRCC1777305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carboxin
CAS 5234-68-4
Molecular Formula C12 H13 N O2 S
Quantity 5 g
Synonym 1,4-Oxathiin-3-carboxanilide, 5,6-dihydro-2-methyl- (7CI,8CI), 5,6-Dihydro-2-methyl-N-phenyl-1,4-oxathiin-3-carboxamide, 1-Oxa-2-methyl-3-(anilidocarbonyl)-4-thiacyclohex-2-ene, 2,3-Dihydro-5-carboxanilido-6-methyl-1,4-oxathiin, 2,3-Dihydro-6-methyl-1,4-oxathiin-5-carboxanilide, 2,3-Dihydro-6-methyl-1,4-oxathiine-5-carboxylic acid anilide, 2,3-Dihydro-6-methyloxathiin-5-carboxanilide, 5,6-Dihydro-2-methyl-1,4-oxathiin-3-carboxanilide, Carbathiin, Carboxin, Carboxine, D 735, DCMO, DMOC, F 735, Fenoxan, Karboxyn, N-Phenyl-2-methyl-5,6-dihydrooxathiin-3-carboxamide, NSC 263492, V 4X, Vitaflo 250, Vitavax, Vitavax 100, Vitavax 34, Vitavax 735D, Vitavax 75W, Vitavax EC 200
InChI Formula InChI=1S/C12H13NO2S/c1-9-11(16-8-7-15-9)12(14)13-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3,(H,13,14)
SMILES CC1=C(SCCO1)C(=O)Nc2ccccc2
IUPAC Name 6-methyl-N-phenyl-2,3-dihydro-1,4-oxathiine-5-carboxamide
Molecular Weight (g/mol) 235.3
Formula Weight 235.0667
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.