missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-hydroxy-3-methyl-4H-chromen-4-one, TRC

Product Code. 30356561
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356561 50 mg -
30349900 500 mg -
30362463 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30356561 Supplier Toronto Research Chemicals Supplier No. TRCC58825050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197504-51-1
Molecular Formula C10H7O3Cl
Molecular Weight (g/mol) 210.61
IUPAC Name 6-chloro-4-hydroxy-3-methylchromen-2-one
SMILES CC1=C(O)c2cc(Cl)ccc2OC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H7ClO3/c1-5-9(12)7-4-6(11)2-3-8(7)14-10(5)13/h2-4,12H,1H3
Formula Weight 210.0084
Chemical Name or Material 6-Chloro-2-hydroxy-3-methyl-4H-chromen-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.