missing translation for 'onlineSavingsMsg'
Learn More

D-Glucuronic Acid 3-Phenylpropyl Ester, TRC

Product Code. 30357163
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357163 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357163 Supplier Toronto Research Chemicals Supplier No. TRCD20703025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Glucuronic Acid 3-Phenylpropyl Ester
CAS 1701402-25-6
Molecular Formula C15 H20 O7
Quantity 25 mg
Synonym 3-Phenylpropyl (2S,3S,4S,5R)-2,3,4,5-Tetrahydroxy-6-oxohexanoate
InChI Formula InChI=1S/C15H20O7/c16-9-11(17)12(18)13(19)14(20)15(21)22-8-4-7-10-5-2-1-3-6-10/h1-3,5-6,9,11-14,17-20H,4,7-8H2/t11-,12+,13-,14-/m0/s1
SMILES O[C@@H](C=O)[C@@H](O)[C@H](O)[C@H](O)C(=O)OCCCc1ccccc1
IUPAC Name 3-phenylpropyl (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate
Molecular Weight (g/mol) 312.315
Formula Weight 312.121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.