missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorambucil, TRC
Specifications
Specifications
| Chemical Name or Material | Chlorambucil |
| CAS | 305-03-3 |
| Molecular Formula | C14 H19 Cl2 N O2 |
| Quantity | 25 g |
| Synonym | Benzenebutanoic acid, 4-[bis(2-chloroethyl)amino]-, Butyric acid, 4-[p-[bis(2-chloroethyl)amino]phenyl]- (8CI), 4-[Bis(2-chloroethyl)amino]benzenebutanoic acid, 4-[p-[Bis(2-chloroethyl)amino]phenyl]butyric acid, Ambochlorin, Amboclorin, CB 1348, Chlorambucil, Chloraminophene, Chlorbutin, Chlorobutine, Ecloril, Leukeran, Leukeran Tablets, Linfolizin, Linfolysin, Lympholysin, NCI 3088, NSC 3088, γ-[p-Bis(2-chloroethyl)aminophenyl]butyric acid, γ-[p-Di(2-chloroethyl)aminophenyl]butyric acid |
| InChI Formula | InChI=1S/C14H19Cl2NO2/c15-8-10-17(11-9-16)13-6-4-12(5-7-13)2-1-3-14(18)19/h4-7H,1-3,8-11H2,(H,18,19) |
| SMILES | OC(=O)CCCc1ccc(cc1)N(CCCl)CCCl |
| IUPAC Name | 4-[4-[bis(2-chloroethyl)amino]phenyl]butanoic acid |
| Molecular Weight (g/mol) | 304.21 |
| Formula Weight | 303.0793 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?