missing translation for 'onlineSavingsMsg'
Learn More

p-Chlorophenylguanidine Hydrochloride, TRC

Product Code. 30357654
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357654 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357654 Supplier Toronto Research Chemicals Supplier No. TRCC37774025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Chlorophenyl)guanidine Hydrochloride
CAS 14279-91-5
Molecular Formula C7 H8 Cl N3 . Cl H
Quantity 25 mg
Synonym (4-Chlorophenyl)guanidine Hydrochloride, Guanidine, N-(4-chlorophenyl)-, hydrochloride (1:1), Guanidine, (4-chlorophenyl)-, monohydrochloride (9CI), Guanidine, (p-chlorophenyl)-, monohydrochloride (8CI), 4-Chlorophenylguanidine hydrochloride, BW 65-100, p-Chlorophenyl urea hydrochloride
InChI Formula InChI=1S/C7H8ClN3.ClH/c8-5-1-3-6(4-2-5)11-7(9)10;/h1-4H,(H4,9,10,11);1H
SMILES Cl.NC(=N)Nc1ccc(Cl)cc1
IUPAC Name 1-(4-chlorophenyl)guanidine;hydrochloride
Molecular Weight (g/mol) 206.07
Formula Weight 205.0174
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.