missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cysteinylacetaminophen-D5 (major), Trifluoroacetic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Cysteinylacetaminophen-D5 Trifluoroacetic Acid Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | C11H9D5N2O4S•C2HF3O2 |
| Quantity | 10 mg |
| Synonym | S[5-(Acetylamino)-2-hydroxyphenyl]-L-cysteine-d5 (major) Trifluoroacetic Acid Salt, APAP-CYS-D5 (major), trifluoroacetic acid salt |
| InChI Formula | InChI=1S/C11H14N2O4S.C2HF3O2/c1-6(14)13-7-2-3-9(15)10(4-7)18-5-8(12)11(16)17;3-2(4,5)1(6)7/h2-4,8,15H,5,12H2,1H3,(H,13,14)(H,16,17);(H,6,7)/t8-;/m0./s1/i1D3,2D,4D; |
| SMILES | OC1=CC([2H])=C(NC(C([2H])([2H])[2H])=O)C([2H])=C1SC[C@H](N)C(O)=O.FC(F)(C(O)=O)F |
| IUPAC Name | S-(5-(acetamido-2,2,2-d3)-2-hydroxyphenyl-4,6-d2)-L-cysteine 2,2,2-trifluoroacetic acid |
| Molecular Weight (g/mol) | 389.36 |
| Formula Weight | 389.0917 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?