missing translation for 'onlineSavingsMsg'
Learn More

Chromium Picolinate, TRC

Product Code. 30358886
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358886 2.5 g 2.5g
30356487 25 g -
30368763 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358886 Supplier Toronto Research Chemicals Supplier No. TRCC4327452.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14639-25-9
Molecular Formula C18 H12 Cr N3 O6
Molecular Weight (g/mol) 418.3
Synonym Chromium(III) Picolinate, Chromium, tris(2-pyridinecarboxylato-κN1,κO2)- (9CI), 2-Pyridinecarboxylic acid, chromium complex (ZCI), Chromium, tris(2-pyridinecarboxylato-N1,O2)- (ZCI), Chromium, tris(picolinato)- (7CI, 8CI), 2-Pyridinecarboxylic acid, chromium(3+) salt, 2-Pyridinecarboxylic acid, chromium(3+) salt (3:1), Chromax, Chromium picolinate, Chromium(3+) 2-pyridinecarboxylate, Chromium(III) picolinate, Chromium(III) trispicolinate, CP 200, Tris(picolinato)chromium
IUPAC Name chromium(3+);pyridine-2-carboxylate
SMILES O=C1C2=N(C=CC=C2)[Cr+3]34(N5=C(C=CC=C5)C(=O)[O-]3)(N6=C(C=CC=C6)C(=O)[O-]4)[O-]1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/3C6H5NO2.Cr/c3*8-6(9)5-3-1-2-4-7-5;/h3*1-4H,(H,8,9);/q;;;+3/p-3
Formula Weight 418.0131
Chemical Name or Material Chromium(III) picolinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.